Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds without Caps
Amber glass protects light-sensitive solutions
Brand: DWK Life Sciences W216867
1348.69 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
1382.41 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Type III soda-lime glass
Specifications
4 oz., 125 mL | |
4 oz. | |
1.88 in. | |
White | |
4.4 in. | |
Polypropylene | |
48 mm |
125 mL | |
Amber | |
Soda Lime Glass | |
112 mm | |
Poly/Vinyl | |
22-400 mm | |
Screw Cap |