Learn More
DWK Life Sciences Wheaton™ AC Medium Rounds: With White Polypropylene Cap with Polyvinyl Liner
Ideal for use as sample bottles
710.00 NOK - 2065.00 NOK
Specifications
Color | Clear |
---|---|
Material | Soda Lime Glass |
Closure Color | White |
Closure Liner | Poly/Vinyl |
Closure Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10063823
|
DWK Life Sciences
W216995 |
30 mL | 33-400 mm | ||||||
12369889
|
DWK Life Sciences
W216996 |
60 mL | 38-400 mm |
863.00 NOK
Pack of 48 |
Estimated Shipment: 14-01-2026
Log in to see stock. |
||||
12379889
|
DWK Life Sciences
W216997 |
125 mL | 48-400 mm |
2065.00 NOK
Pack of 24 |
Estimated Shipment: 21-11-2025
Log in to see stock. |
||||
12389889
|
DWK Life Sciences
W216998 |
250 mL | 58-400 mm |
710.00 NOK
Pack of 24 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||
12399889
|
DWK Life Sciences
W216999 |
500 mL | 70-400 mm |
831.00 NOK
Pack of 24 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||
Description
- Manufactured from Wheaton 800 glass, which meets USP Type III standards
- Useful for dry powders, soil samples, and specimen storage
Specifications
Clear | |
White | |
Polypropylene |
Soda Lime Glass | |
Poly/Vinyl | |
Screw Cap |