Learn More
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories
Choose from a wide range of filtration assemblies and accessories to suit your application needs
Brand:  DWK Life Sciences 419331
738.20 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
798.10 NOK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Product Code. 12471571
  • 1185.00 NOK / Each
Genomics Powerhouse
Use promo code 24117 at checkout
View Available Promotions
Only null left
Estimated Shipment: 13-08-2025
Specifications
25mm filtration assembly
Funnel