Learn More
Grant Instruments Well Plate Adaptor
Centrifuge rotor adapter for well plates.
1580.37 NOK
Specifications
Material | Plastic |
---|---|
Autoclavable | Autoclavable |
For Use With (Equipment) | Grant Instruments Centrifuges |
Color | White |
Max. RCF | 1,250 x g |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
30274371
|
Grant Instruments
AP-384 |
348-well PCR Plate Adapter |
1580.37 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30274321
|
Grant Instruments
AP-96 |
96-well PCR Plate Adapter |
1580.37 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Set of 2 adaptors
- To be used with R-2 and Ri-2 rotors
- Rotors can be used with LMC-56, LMC-3000 and LMC-4200R
- Adaptors available for 96-well plates or 384-well plates
Specifications
Plastic | |
Grant Instruments Centrifuges | |
1,250 x g | |
124 mm | |
86 mm |
Autoclavable | |
White | |
2,000 rpm | |
14 mm | |
2/Pk. |