Learn More
Sartorius™ Weight Fork
POA - 1750.00 DKK
Specifications
For Use With (Application) | Prevent any damage to the surface of weights |
---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10042141
|
Sartorius™
YAW42 |
1kg Weight |
1750.00 DKK
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
10534181
|
Sartorius™
YAW43 |
2kg Weight |
POA
Each |
N/A | |||||
10134565
|
Sartorius™
YAW41 |
500g Weight |
POA
Each |
N/A | |||||
Description
Specifications
Prevent any damage to the surface of weights |