Learn More
Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure
Product Code. 15885398 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
Packaging:
Nalgene Bottle
Unit Size:
1 L
100mL
25mL
500mL
CAS: 7732-18-5 | H2O | 18.015 g/mol
Chemical Identifiers
CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
Specifications
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Bottle
Density 1
Quantity 100 mL
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water