Learn More
Wall Cabinet
Brand:  N/A 88V793G
Product Code. 12973445
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The range of Viewcab sheet steel storage cabinets are suitable for a number of working environments including laboratories, schools, universities, factories etc. The cabinets are available in three different sizes. Viewcab doors are reinforced and use 6mm laminated safety glass to BS 6206, so you can view your stored items without compromising security. Chrome lever locking handle with semi-concealed internal mechanism. Steel shelves capacity 75kg per shelf UDL. Laminated glass shelves 50kg per shelf UDL.
Specifications
Specifications
Steel
Wall Mounted
2
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Wall Cabinet > Dimensions (D x W x H): 915 x 300 x 712mm