Learn More
Lovibond™ W600 Borosilicate Glasses
114.00€ - 131.00€
Specifications
Type | W600 Cuvette |
---|---|
Material | Borosilicate Glass |
Quantity | 1Each |
For Use With (Equipment) | Model Fs, PFXi/PFX195, 880, 950 and 995 instruments |
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
11917765
|
Lovibond™
656200 |
50 mm |
131.00€
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
11907765
|
Lovibond™
656020 |
40 mm |
114.00€
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
|||||
Specifications
W600 Cuvette | |
Borosilicate Glass | |
1Each | |
Model Fs, PFXi/PFX195, 880, 950 and 995 instruments |