missing translation for 'onlineSavingsMsg'
Learn More

Vinyltriethylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11303176
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1112-54-5 | C8H18Si | 142.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-54-5
Molecular Formula C8H18Si
Molecular Weight (g/mol) 142.32
MDL Number MFCD00041345
InChI Key HBWGDHDXAMFADB-UHFFFAOYSA-N
Synonym vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane #
PubChem CID 559195
IUPAC Name ethenyl(triethyl)silane
SMILES CC[Si](CC)(CC)C=C

Specifications

Boiling Point 145°C to 146°C
Quantity 5 g
UN Number UN1993
Beilstein 1743830
Flash Point 25°C (77°F)
Formula Weight 142.32
Percent Purity 97%
Refractive Index 1.434
Density 0.771
Chemical Name or Material Vinyltriethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.