Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Useful where conventional filter papers would be destroyed by acids and alkalis
Brand:  Quickfit VB30/08M
 View more versions of this product
Product Code. 15728127
  • 8540.00 NOK / Pack of 5
Only null left
Estimated Shipment: 14-10-2025
Description
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 mL
50 mm
Disc: 30mm
Vacuum Filtration Funnel
Borosilicate Glass
19/26
4