Learn More
Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)
Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.
Brand:  Applied Biosystems™ A31483
 View more versions of this product
Product Code. 15612436
  • 1778.00 NOK / Each
Explore more special offers
Description
Description
Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.
For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.
In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3-m connector cable (flexible)
0.0 to 105.0°C
Automated Thermal Cycler (ATC)
17.9 x 31.7 x 13.3 cm (Block Module)
Connector Cable
Applied Biosystems Automated Thermal Cyclers
1 cable
9.4 kg
±0.25°C (35°C to 99.9°C)
  • ATC block module
  • ATC control module
  • Dell E6540 RC Laptop computer
  • 1 meter connection cable
  • CAT 5 ethernet cable – Store at room temp
<0.5 °C (20 s after reaching 95°C)
Connectivity-LAN
27 Month Warranty
Cabling Option - 3MPlate Format - 96-wellData Connectivity - LANBlock Module Dimensions - 17.9 x 31.7 x 13.3 cmControl Module Dimensions - 25.7 x 33.1 x 7.0 cmIncludes - 3 meter connection cable for Automated Thermal Cycler (ATC)
10-100 μL
17.9 x 31.7 x 13.3 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.