Learn More
Fisherbrand™ Polyethersulfone Membranes
Fisherbrand™ Polyethersulfone Membranes features highly asymmetrical pore structure that offers effective loading capacity and high flow rate.
Brand: Fisherbrand™ 0444A00022
475.00 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Highly asymmetrical pore structure offers effective loading capacity and high flow rate
- Hydrophilic and made with polyethersulfone polymer
- Designed to remove particulates during general filtration
- Low protein and drug binding characteristics make it ideally suited for use in life science applications
Specifications
Yes | |
White | |
0.45 μm | |
21mL/min./cm2 | |
16L/min./cm2 | |
100 Pack | |
Yes |
2.7bar | |
PES | |
Non-sterile | |
Filtration of special chemical reagent, high temperature liquids, filtration of special chemical reagent, high temperature liquids, filtration of drugs | |
Hydrophilic | |
Plain Disc | |
25 mm |