Learn More
Thermo Scientific™ Nunc™ Conical Sterile Polypropylene Centrifuge Tubes
GREENER_CHOICE
These premium, high-quality conical tubes are environmentally friendly, and allow for increased traceability with the largest writing area on the market.
Brand:  Thermo Scientific™ 339651
778.03 NOK valid until 2025-12-31
Use promo code "24071" to get your promotional price.
 View more versions of this product
Product Code. 11307211
  • 3310.75 NOK / Pack of 500
In Stock
Edge
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Nunc Conical Sterile Polypropylene Centrifuge Tubes are premium, high-quality conical tubes that are environmentally friendly, offer the highest cleanliness with a recyclable plastic rack and allow for increased traceability with the largest writing area on the market. Nunc Labware Products are made from high-purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features
  • Nonpyrogenic: Endotoxin tested (0.5 EU/mL)
  • Non-Cytotoxic
  • Biologically inert: USP biological reactivity class VI
  • High level of sterility: SAL 10-6
  • RNase/DNase-free
  • Leak-proof to protect your samples
  • Made from high-purity resins and molded using state-of-the-art processes; safe alternative to glass without sacrificing accuracy
  • Racked product includes environmentally friendly recyclable plastic rack that reduces landfill waste compared to the traditional foam packaging
  • Plug sealed cap
  • Tube, cap and rack made of virgin resin
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15 mL
Reusable and recyclable polypropylene rack
25
25 Pk.
Sustainable packaging
Conical
10,500 x g
Polypropylene
500/Cs.
Racked
Sterile
Cylindrical
Centrifuge Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.