Learn More
Corning™ CellBIND™ Surface Microplates
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
Brand: Corning™ 3335
2526.23 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Certifications
Certified nonpyrogenic
Specifications
Clear | |
Polystyrene | |
2.9 mL (Well) | |
9.5 cm2 | |
Rectangular | |
CellBIND | |
1.9 to 2.9 mL | |
Lid | |
5/Pk. |
Yes | |
Flat | |
Clear | |
Polystyrene | |
Sterile | |
Multi-well Cell Culture Plate | |
6 | |
50/Cs. |