Learn More
Cytiva VacuCap™ 90 PF Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
2435.00 NOK - 3140.00 NOK
Specifications
Material (Membrane) | Supor (Hydrophilic PES) |
---|---|
Pore Size | 0.8/0.2 μm |
Sterility | Sterile |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
17154371
|
Cytiva
4628 |
90 mm |
3140.00 NOK
Pack of 10 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
|||||
17194371
|
Cytiva
4638 |
60 mm |
2435.00 NOK
Pack of 10 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
0.8/0.2 μm | |
Sterile |