Learn More
UVP UV lamp UVM-26 EL hand held 220V 50Hz 6W 302 nm UVP
Brand:  UVP 95-0276-02
5867.57 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11748251
  • 6990.00 NOK / Each
Only null left
Estimated Shipment: 06-10-2025