Learn More
Vilber Lourmat™ UV Lamp Stand
Rich and robust table stand for ultraviolet lamps 4/8 W.
Brand: Vilber Lourmat™ 9130-2003-1
1183.38 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Stand for UV lamps 4/8W
- Double wavelength lamp
- 254/365 nm Intensity
- Lamp 2 x 6 W
- M, 2 table stand
Specifications
UV Lamp Stand | |
Stand for UV lamps 4/8W | |
4/8w UV lamp |