Learn More
Emballator UN Approved Cans
Made of HDPE. Emballator UN Approved Cans approved by NET for storage and transport of dangerous goods.
Brand:  Emballator 48H0517-410-300
Encompass
 View more versions of this product
Product Code. 11394405
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Material: HDPE, approved for food contact
  • Color: Nature
  • UN Approved tamper evident closure
  • Approved by NET for storage and transport of dangerous goods
Specifications
Specifications
UN Approved Can
Natural
190 mm
296 mm
175 g
5 L
HDPE
130 mm
40 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Emballator UN Approved Cans