Learn More
UBQLN2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-23449-20UL
 View more versions of this product
Product Code. 18087405
  • 1696.32 NOK / 20µL
Description
The UBQLN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBQLN2. This antibody recognizes human, rat antigen. The UBQLN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
UBQLN2
1000 μg/mL
CHAP1, CHAP1/DSK2, Dsk2, DSK2 homolog, hPLIC 2, HRIHFB2157, LIC 2, N4BP4, PLIC 2, PLIC2, RIHFB2157, ubiquilin 2, UBQLN2
20 μL
29978
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Rat
150 kDa