Learn More
UBQLN2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-23449-100UL
Description
The UBQLN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBQLN2. This antibody recognizes human, rat antigen. The UBQLN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
UBQLN2 | |
1000 μg/mL | |
CHAP1, CHAP1/DSK2, Dsk2, DSK2 homolog, hPLIC 2, HRIHFB2157, LIC 2, N4BP4, PLIC 2, PLIC2, RIHFB2157, ubiquilin 2, UBQLN2 | |
100 μL | |
29978 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Immunofluorescence | |
CoraLite Plus 488 | |
Rabbit | |
Primary | |
Human, Rat | |
150 kDa | |