Learn More
DuPont™ Tyvek™ IsoClean™ Slip-retardant Shoe Cover
Made of high density polyethylene, Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
1705.00 NOK
Specifications
Color | White |
---|---|
Sterility | Non-sterile |
Model | IC451S00 |
Seam Type | Serged |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16428363
|
DuPont™
D15531632 |
Large |
1705.00 NOK
Pack of 100 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
16438363
|
DuPont™
D15531633 |
Medium |
1705.00 NOK
Pack of 100 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
Description
- Gripper™ soles provide enhanced slip-retardance and robustness
- Delivers an ideal balance of protection, durability and comfort li>Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms
Specifications
White | |
IC451S00 | |
(EU) 2016/425, EN 14126 |
Non-sterile | |
Serged |