Learn More
Ismatec™ Tygon™ MHSL 2001 3-stop Tubing
Transparent, plasticizer-free 3-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.
Brand: Ismatec™ SC0810
This item is not returnable.
View return policy
Description
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated.
- Applications
- Acids: Excellent
- Alkaline solutions: Excellent
- Solvents: Good/Excellent
- Complies with: FDA.
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
- Note: Although designed for MS/CA cassettes, this tubing has only 2 stoppers!
Specifications
FDA | |
Autoclavable | |
Tygon MHSL 2001 | |
30 cm | |
peristaltic pump | |
-73°C | |
Acids, Alkalies | |
CO2 1140, O2 76, N2 190 | |
57°C |
Color-code: Purple-Purple | |
2.06 mm | |
3.76 mm | |
0.85 mm | |
For MEK and other aggressive solvents. MS/CA cassettes | |
6 pcs./Pk. | |
2.06mm I.D., 3.76mm O.D. | |
Transparent | |
2-stop Tubing |