Learn More
Labplas Twirl'EM™ Clear Polyethylene Sampling Bags
Collect solid, semi-solid or liquid samples in the laboratory using Twirl'EM™ Clear Polyethylene Sampling Bags. The bags feature a twirl round metal wire closure system with safety tabs to provide a leakproof seal.
1862.14 NOK - 6605.00 NOK
Specifications
Color | Clear |
---|---|
Material | Polyethylene |
Sterility | Sterile |
Quantity | 1000 Pk. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11582773
|
Labplas
EFL-1012 |
2250 mL |
6300.00 NOK
Pack of 1000 |
Estimated Shipment: 31-10-2025
Log in to see stock. |
|||||
11592773
|
Labplas
EFL-1015-FS-1 |
4000 mL |
6605.00 NOK
Pack of 1000 |
Estimated Shipment: 28-10-2025
Log in to see stock. |
|||||
11522773
|
Labplas
EPL-3070-FS-1_OLD |
150 mL |
1862.14 NOK
Pack of 1000 |
Estimated Shipment: 12-11-2025
Log in to see stock. |
|||||
11552773
|
Labplas
EPL-4512-FS-1 |
800 mL |
2795.00 NOK
Pack of 1000 |
Estimated Shipment: 28-10-2025
Log in to see stock. |
|||||
11542773
|
Labplas
EPL-5590-FS-1 |
650 mL |
3170.00 NOK
Pack of 1000 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
11562773
|
Labplas
EPL-4515-FS-1 |
1060 mL |
3125.00 NOK
Pack of 1000 |
Estimated Shipment: 25-11-2025
Log in to see stock. |
|||||
11572773
|
Labplas
EPL-5515-FS-1 |
1240 mL |
3390.00 NOK
Pack of 1000 |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||||
Description
- Sterile
- Clear polyethylene
- Without side seals, opening at width
- Pull tabs to open bag without touching edges
- Twirl round metal wire closure system with safety tabs
Specifications
Clear | |
Polyethylene | |
Sterile | |
1000 Pk. |