Learn More
Tubing Holder
Polypropylene clip captures and holds flexible tubing firmly in place in beakers or other open containers.
Brand: Bel-Art - SP Scienceware F18235-0000
This item is not returnable.
View return policy
Description
- Fits snugly over edge of beaker
- Long arm measures 2 in. (51mm) with hole to accept tubing up to 0.375 in. (9.5mm) O.D.
- Autoclavable
Specifications
Autoclavable | |
Polypropylene | |
Tubing Holder |