Learn More
Thermo Scientific Chemicals Tropaeolin O, pure
Product Code. p-209866 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym Acid orange 6, C.I. 14270, 4-(2, 4-Dihydroxyphenylazo)benzenesulfonic acid, sodium salt
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]
Specifications
Beilstein 16, 275
Merck Index 15, 9954
Solubility Information Solubility in water: Soluble. Other solubilities: Soluble in alcohol
Infrared Spectrum Authentic
Formula Weight 316.26
Grade Extra Pure
Packaging Glass bottle
Visual Transition Interval from pH 11.1 (yellow) to pH 12.7 (orange)
Color Brown to Rust
Quantity 25 g
Physical Form Powder
Chemical Name or Material Tropaeolin O