Learn More
Tributylborane, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Product Code. 10731345
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-56-5, 109-99-9
Molecular Formula C12H27B
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009423
InChI Key CMHHITPYCHHOGT-UHFFFAOYSA-N
Synonym tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane
PubChem CID 31216
IUPAC Name tributylborane
SMILES CCCCB(CCCC)CCCC
Specifications
Color Colorless to Yellow
Linear Formula [CH3(CH2)3]3B
Concentration 0.94 to 1.06M
Fieser 10,410
Density 0.8470g/mL
Formula Weight 182.16
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.847
Quantity 100 mL
Flash Point −20°C
Physical Form Solution
Chemical Name or Material Tributylborane, 1M Solution in Tetrahydrofuran