Learn More
Tri-n-butyl phosphite, 94%
CAS: 102-85-2 | C12H27O3P | 250.319 g/mol
763.00 NOK - 2495.00 NOK
Chemical Identifiers
CAS 102-85-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00009437
InChI Key XTTGYFREQJCEML-UHFFFAOYSA-N
Synonym tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
PubChem CID 7623
IUPAC Name tributyl phosphite
SMILES CCCCOP(OCCCC)OCCCC
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353456
SDS View Documents
Thermo Scientific Alfa Aesar
L14307.AE
100 mL
763.00 NOK
100mL
11363456
SDS View Documents
Thermo Scientific Alfa Aesar
L14307.AP
500 mL
2495.00 NOK
500mL
Description
Description
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.
Solubility
Not miscible or difficult to mix in water.
Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.
Specifications
Chemical Identifiers
102-85-2
250.319
XTTGYFREQJCEML-UHFFFAOYSA-N
7623
CCCCOP(OCCCC)OCCCC
C12H27O3P
MFCD00009437
tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
tributyl phosphite
Specifications
102-85-2
0.915
82°C (179°F)
1.432
1703866
tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
XTTGYFREQJCEML-UHFFFAOYSA-N
tributyl phosphite
7623
94%
-80°C
125°C to 128°C (10 mmHg)
C12H27O3P
MFCD00009437
Air and moisture sensitive
Not miscible or difficult to mix in water.
CCCCOP(OCCCC)OCCCC
250.319
250.32
Tri-n-butyl phosphite
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H312-H315-H319-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H312+H332-H315-H319
missing translation for 'einecsNumber' : 203-061-3
missing translation for 'rtecsNumber' : TH0875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen