missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-3 siRNA
Blocks transcription and translation of Caspase-3 from Human.
Brand: enQuireBio™ QX40-10nmol
This item is not returnable.
View return policy
Specifications
NP_004337 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
836 | |
10 nmol | |
Human |
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. | |
CASP3 | |
siRNA | |
SCA-1, CPP32, CPP32B CASP3 | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction