Learn More
enQuireBio™ TranslationBlocker Human Apoptosis regulator BAX siRNA
Blocks transcription and translation of Apoptosis regulator BAX from Human.
Brand:  enQuireBio™ QX6-2nmol
 View more versions of this product
Product Code. 16079251
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_004315
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
581
2 nmol
Human
KAT-4 Human Thyroid Carcinoma Cells and Human T Cells.
BAX
siRNA
BCL2L4
Lyophilized