Learn More
Alla France™ Traceable Mineral Oil Hydrometer
73.00€ - 75.10€
Specifications
Quantity | 1Each |
---|---|
Type | Traceable Mineral Oil Hydrometer |
Accuracy | ±0.1°API |
Length (Metric) | 335 mm |
Product Code | Brand | Specific Gravity Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Specific Gravity Range | Price | Quantity & Availability | |||||
11762565
|
Alla France™
3400FC009/60-qp |
9 to 21° API |
73.00€
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
11782565
|
Alla France™
3400FC029/60-qp |
29 to 41° API |
75.10€
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
Specifications
1Each | |
Traceable Mineral Oil Hydrometer | |
±0.1°API | |
335 mm |