Learn More
BINDER™ Top Access Port (50 mm)
For Use With: FED 56, FED 115, FED 260, FED 400, FED 720
Brand: BINDER™ 8012-1381
7066.06 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use on top
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 50 mm, top | |
Steel | |
Silicone plug |
BINDER Chambers Series FED | |
Access Port | |
50 mm |