Learn More
Terumo™ Three-part Syringes
Click to view available options
Unit Size:
Pack of 100
Description
- Highly transparent barrel for easy recognition of air bubbles
- Clear printing for precise dosage management
- Latex-free syringes contain no natural rubber
- Environmental-friendly Designed to have a minimum of waste
Specifications
Specifications
Certifications/Compliance | Class IIa, sterile product following MDD 93/42/EEC |
Material | Polypropylene, Silicone Oil (Lubricant), TPE |
Color | Clear |
For Use With (Application) | Injection of fluids - immediately after filling - into the human body, for aspiration of fluids or for the sampling of blood or other body fluids |
Latex Free | Yes |
Product Type | 3-part Syringe |
Disposable | Single-use |
Graduations | 0.2 mL |
Volume (Metric) | 5 mL |
Needle Point Style | Concentric |