Learn More
Terumo™ Three-Part Syringe
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 100
Description
- Ultra-sharp needles
- Clear barrels
- Bold, precise scale markings
- Smooth plunger action
- Tamper evident peel packs
Specifications
Specifications
Autoclavable | Autoclavable |
Material | Polypropylene |
Needle Gauge | 21 G |
Latex Free | Yes |
Length (Metric) | 40 mm |
Product Type | 3-part Syringe |
Quantity | 100/Pk. |
Volume (Metric) | 10 mL |
Sterility | Sterile |