Learn More
Hanna Instruments™ Thermocouple Probe Hi-766Pd
Brand:  Hanna Instruments™ HI766PD
935.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10418642
  • 1605.00 NOK / Each
Only null left
Estimated Shipment: 29-08-2025
Description
Interchangeable K type stainless steel 250mm x 3mm diameter 300°C max air temperature Hanna