Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
This Quick Column Change accessory is compatible with Agilent Instrumentation
46.55€ - 86.70€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13406956
|
Thermo Scientific™
60180-872 |
S/SL Injector Adaptor | N/A | ||||||
13496956
|
Thermo Scientific™
60180-878 |
TCD Detector Adaptor | N/A | ||||||
13446956
|
Thermo Scientific™
60180-873 |
Tight Nut | N/A | ||||||
Description
QCC (Quick Column Change)-Leak Free Column Installation
- Tool-free installation of analytical GC column
- Graphite ferrules
Complete QCC Kits Includes:
- Injector Adaptor
- Detector Adaptor
- Tight Nut
- Ferrules (1 each 0.25, 0.32,0.53)
- Ceramic Column Cutter