Learn More
Memmert™ Temperature Restriction for UF260
Optional temperature restriction between 60°C and 250°C.
Brand: Memmert™ UF260A8
160.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- 1 Choice
- Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
UF260 Oven | |
Temperature Restriction |