Learn More
BRAND™ Filling Tube for Dispensette™ S / S Organic
Manufactured from FEP, these Telescoping Filling Tubes adjust to various bottle heights.
Brand: BRAND™ 708218
257.95 NOK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
This item is not returnable.
View return policy
Description
- FEP
Specifications
FEP | |
1/Pk. | |
7.6 mm |
25, 50, 100mL nominal capacity | |
From 170 to 330 mm |