Learn More
TBL1X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13540-1-AP-20UL
 View more versions of this product
Product Code. 16861804
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroid receptors) complex along with histone deacetylase 3 protein. This gene is located adjacent to the ocular albinism gene and it is thought to be involved in the pathogenesis of the ocular albinism with late-onset sensorineural deafness phenotype. Four transcript variants encoding two different isoforms have been found for this gene. This gene is highly similar to the Y chromosome TBL1Y gene.
Specifications
Specifications
TBL1X
Polyclonal
Unconjugated
TBL1X
EBI, SMAP55, TBL1, TBL1X
Rabbit
Antigen Affinity Chromatography
RUO
21372, 302711, 6907
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60907, Q9QXE7
TBL1X
TBL1X Fusion Protein Ag4477
20 μL
Primary
Mouse, Human, Rat