Learn More
Buerkle™ T-Piece with Inner Thread
Manufactured from polypropylene.
61.53 NOK - 102.27 NOK
Specifications
Certifications/Compliance | EU Foodstuffs |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Type | T-Piece Inner Thread |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
10396152
|
Buerkle™
8525-0012 |
17.5 mm |
61.53 NOK
Each |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
10269210
|
Buerkle™
8525-0034 |
23 mm |
83.68 NOK
Each |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
10692862
|
Buerkle™
8525-0100 |
28.5 mm |
102.27 NOK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
EU Foodstuffs | |
Autoclavable | |
Polypropylene | |
T-Piece Inner Thread |