Learn More
Hamilton™ Terasaki Dispenser
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
Brand:  Hamilton™ 201910
12564.49 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10251822
  • 12880.00 NOK / Each
Description
  • Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
  • Uses removable needles
  • Available for a range of volumes
Specifications
Specifications
1/Pk.
Terasaki Dispenser
50 μL