Learn More
Sigma Laborzentrifugen™ Swing-out Rotors for Sigma™ Series 2 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Swing-out Rotors for Sigma™ Series 2 Centrifuges are ideal for use in medical and research laboratories.
708.00€ - 3050.00€
Product Code | Brand | Capacity | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | For Use With (Equipment) | Price | Quantity & Availability | ||||
10036111
|
Sigma Laborzentrifugen™
11030 |
120 mL | Sigma 2-6, 2-6E, Buckets: 13031, 13033, 13034, 13035, 13037 |
708.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
11796698
|
Sigma Laborzentrifugen™
11409 |
24 x 1.5 x 75 mm | Sigma 2-16 and 2-16KL |
1125.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10779212
|
Sigma Laborzentrifugen™
11124 |
24 x 1.5/2.2 mL | Sigma 2-16, 2-16P, 2-16PK, 2-16K, 3-18K, 3-18KH |
1940.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10289671
|
Sigma Laborzentrifugen™
11121 |
Sigma 2-6, 4-16, 4-16K, 4-16KH, Microtiter Plates |
2245.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
10525902
|
Sigma Laborzentrifugen™
11123 |
2 to 6 Microtiter Plates | Sigma 2-16, 2-16K, 2-16KCH, 4-16, 4-16K, 4-16KH, 4-16KCH |
3050.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
- Swing-bucket rotors for sedimentation with smaller centrifugal forces of up to approximately 6000 x g
- A higher yield may be obtained as it is possible to fill the centrifuge test tube completely
- Sedimentation takes place in the middle of the bottom of the test tube enabling easy separation of the phases
- Sigma supplies matching fixed-angle, swing-out and drum rotors for all Sigma centrifuges
- Sigma rotors are precision-made from modern, certified high-performance materials with traceable batch numbers
- Thanks to many years of experience and close contact with our customers, we are able to offer an extensive selection of rotors
- Sigma rotors are characterized by extra capacity or higher maximum speed
- Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Swing-out Rotor |