Learn More
Karl Hecht™ Assistent™ SuperFrost™ Plus Microscope Slides
Attracts electrostatically fresh frozen tissue section and cytology preparations, binding them to the slides. Karl Hecht™ Assistent™ SuperFrost™ Plus Microscope Slides are recommended for anatomic and molecular pathology laboratories.
Brand: Karl Hecht™ 42409110
This item is not returnable.
View return policy
Description
- Permanent positive charge placed upon slides
- Proven to be superior for cell adhesion in cytocentrifuge preparations in standard Papanicolaou smears
- Preparations can be marked and identified by the coated labeling field
- Coating of labeling field is resistant to common solvents
- Thickness of approx. 1mm
Specifications
Yes | |
White | |
Solvent Resistant |