Learn More
Thermo Scientific™ SUN-SRi™ Dry Vials™
Nitrogen-purged vials used for low-level water determination in the tobacco industry
Brand:  Thermo Scientific™ 2001/14
 View more versions of this product
Product Code. 12054598
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »
Description
Description
  • Standard-opening, 1.5mL, 12 × 32mm vials for use in FTC regulation Volume 32 #147 and Health Canada Official Method T-115 to determine the amount of tar, nicotine, and carbon monoxide in tobacco smoke
  • Nitrogen purged to provide true total particulate matter readings free of contaminating water/water vapor from vial surfaces
  • Includes aluminum seal closures with PTFE/rubber septa
  • Available in Clear, Type 1 Class A or Amber, Type 1 Class B borosilicate glass