Learn More
Hirschmann™ Suction Tube with Recirculation
Screw-coupled suction tube securely attached to the Hirschmann™ liquid handling device. Longer suction tube for remotely placed media reservoir.
Brand: Hirschmann™ 9314006
352.60 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
This item is not returnable.
View return policy
Description
- Screw-coupled
- Safely attached to the instrument
Specifications
Suction Hose | |
Ceramus, Solaris | |
w/ Recirculation | |
2000 mm |