Learn More
Memmert™ Subframe for SF75, Adjustable up to 600 mm
Adjustable subframe for Memmert Sterilizers.
Brand: Memmert™ SF75B29747
3965.89 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adjustable in height
Specifications
Adjustable | |
SF75 Sterilizer | |
Subframe |