Learn More
Memmert™ Subframe on Castors for SF75
Subframe on wheels for Memmert Sterilizers.
Brand: Memmert™ SF75B29748
5872.79 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- On Castors
- Height: 660 mm
Specifications
On Wheels | |
SF75 Sterilizer | |
Subframe |