Learn More
Techne™ Stuart™ Virgin Polypropylene Microtubes
2335.00 NOK - 6355.00 NOK
Specifications
Closure Material | Virgin polypropylene |
---|---|
Color | Clear |
Material | Polypropylene (Virgin) |
No. per Pack | 1000/Pk. |
Packaging | 1000/Pk. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13018513
|
Techne™
99968-00 |
200 μL |
6355.00 NOK
Pack of 1000 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
11573919
|
Techne™
99968-01 |
500 μL |
2335.00 NOK
Pack of 1000 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
Specifications
Virgin polypropylene | |
Polypropylene (Virgin) | |
1000/Pk. |
Clear | |
1000/Pk. |