Learn More
Cole-Parmer™ Stuart™ Shaking Water Bath Stainless Steel Rack
Enhance the versiltility of your Stuart™ water bath
2685.00 NOK - 2715.00 NOK
Specifications
For Use With (Equipment) Shaking Water Bath
Material Stainless Steel
View More Specs
Products 6
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
10725004
View Documents
Cole-Parmer™
12152-95
120 x 13 mm Culture Tubes
2715.00 NOK
Each
Only null left
Estimated Shipment: 25-08-2025
Add to basket
12738988
View Documents
Cole-Parmer™
12152-93
25 x 50 mL Centrifuge Tubes
2685.00 NOK
Each
Only null left
In Stock
Add to basket
10181133
View Documents
Cole-Parmer™
12152-94
143 x 1.5 mL Microtubes
2715.00 NOK
Each
Only null left
Estimated Shipment: 25-08-2025
Add to basket
12728988
View Documents
Cole-Parmer™
12152-98
30 x 26 mm Culture Tubes
2715.00 NOK
Each
Only null left
Estimated Shipment: 25-08-2025
Add to basket
10600394
View Documents
Cole-Parmer™
12152-96
72 x 16 mm Culture Tubes
2715.00 NOK
Each
Only null left
Estimated Shipment: 25-08-2025
Add to basket
12718988
View Documents
Cole-Parmer™
12152-97
56 x 15 mL Centrifuge Tubes
2715.00 NOK
Each
Only null left
Estimated Shipment: 25-08-2025
Add to basket
Description
Description
  • Choose from these accessories to make your unit more versatile
  • Designed to be easily interchanged with the stuart water baths and shaking water baths
Specifications
Specifications
Shaking Water Bath