Learn More
Cole-Parmer™ Stuart™ Heater Block Clamp
Turn your existing 6 x 6 in. hot plate into a heating mantle
Brand:  Cole-Parmer™ 04805-78
Product Code. 15163027
  • 2535.00 NOK / Each
Only null left
Estimated Shipment: 19-09-2025
Description
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
Specifications
Specifications
Block Clamp
clamp SHB250CLAMP
Medium