Learn More
Cole-Parmer™ Stuart™ Gabled Lid
Enhance the performance of your Techne baths and circulators
Brand:  Cole-Parmer™ 12122-97
4601.74 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12767969
  • 5135.00 NOK / Each
Only null left
Estimated Shipment: 24-09-2025
Description
  • Lids, trays, temperature probes, and other accessories for your bath.
  • Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.
Specifications
Specifications
18 and 26L Bath